4-Bromo-5-methyl-3-nitro-1H-pyrazole 97%

Supplier: Ambeed
Warning

A215340-250MG A215340-5G A215340-10G
103818-480EA 17 USD
103818-480 103849-184 76814-708 77303-410 77671-922
4-Bromo-5-methyl-3-nitro-1H-pyrazole 97%
4-Bromo-3-methyl-5-nitro-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₄BrN₃O₂
MW: 206.00 g/mol
Boiling Pt: 331.9±37.0 °C (760 mm Hg)
Flash Pt: 154.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00195045
CAS Number: 70951-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR