4-Bromo-5-methyl-1H-indazole 97%

Supplier: Ambeed
Warning

A243641-1G A243641-5G A243641-10G A243641-250MG
103811-530EA 8 USD
103811-530 103851-352 77090-554 77090-556 77288-790 77288-792 77288-794
4-Bromo-5-methyl-1H-indazole 97%
4-Bromo-5-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
Boiling Pt: 344.6 °C (760 mm Hg)
Flash Pt: 162.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11044156
CAS Number: 926922-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR