4-Bromo-5-methyl-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole 97%

Supplier: Ambeed
Warning

A185511-1G A185511-25G A185511-5G A185511-100MG A185511-10G A185511-250MG
103815-860EA 36 USD
103815-860 103846-428 77084-062 77084-064 77084-066 77138-870 77138-872 77138-874
4-Bromo-5-methyl-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole 97%
4-Bromo-5-methyl-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole

Precaución: for Research Use Only

Formula: C₁₃H₁₅BrN₂O
MW: 295.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19443358
CAS Number: 1926172-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR