4-Bromo-5-methoxy-1H-indole 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A158952-250MG A158952-1G A158952-5G A158952-100MG
103805-202EA 107 USD
103805-202 103843-414 76984-976 77300-382 77300-384 77683-326
4-Bromo-5-methoxy-1H-indole 98%, Ambeed.Inc
4-Bromo-5-methoxy-1H-indole

Precaución: For research use only.

Formula: C₉H₈BrNO
Boiling Pt: 349.2±22.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD12973810
CAS Number: 90858-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR