4-Bromo-5-iodopyridin-2-amine 97%

Supplier: Ambeed
Warning

A195492-100MG A195492-10G A195492-250MG A195492-1G A195492-5G
103816-756EA 53 USD
103816-756 103847-368 76825-370 76825-372 76825-374 77085-314 77085-316
4-Bromo-5-iodopyridin-2-amine 97%
4-Bromo-5-iodopyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄BrIN₂
MW: 298.91 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16658670
CAS Number: 1186115-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR