4-Bromo-5-fluoro-2-nitrophenol 97%

Supplier: Ambeed
Danger

Synonyms: 5-Bromo-4-fluoro-2-hydroxynitrobenzene

A106790-100MG A106790-10G A106790-250MG A106790-1G A106790-5G A106790-25G
103797-076EA 25 USD
103797-076 103822-834 76869-470 76869-472 76869-474 77309-682 77309-684 77610-846
4-Bromo-5-fluoro-2-nitrophenol 97%
4-Bromo-5-fluoro-2-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₃
MW: 235.995 g/mol
Boiling Pt: 266.8±35.0 °C (760 mm Hg)
Flash Pt: 115.2 °C
Storage Temperature: Ambient
MDL Number: MFCD14583144
CAS Number: 1016234-87-9
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR