4-Bromo-5-fluoro-2-methylpyridine 97%
Supplier: Ambeed
|
Warning
|
A119974-100MG
A119974-1G
103799-866EA
73
USD
103799-866
76870-156
77311-974
4-Bromo-5-fluoro-2-methylpyridine 97%
4-Bromo-5-fluoro-2-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₅BrFN MW: 190.013 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18251020 CAS Number: 1211590-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...