4-Bromo-5-fluoro-2-methylbenzoic acid 98%

Supplier: Ambeed
Warning

A102188-100MG A102188-10G A102188-250MG A102188-25G A102188-1G A102188-5G
103796-330EA 14 USD
103796-330 103822-024 76982-214 76982-216 76982-218 76982-220 77308-900 77308-902
4-Bromo-5-fluoro-2-methylbenzoic acid 98%
4-Bromo-5-fluoro-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
Melting Pt: 152…153 °C
Storage Temperature: Ambient
MDL Number: MFCD18917157
CAS Number: 1349715-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR