4-Bromo-5-chloropyridin-2-amine 95%
Supplier: Ambeed
|
Warning
|
A161428-10G
A161428-250MG
A161428-25G
A161428-5G
A161428-1G
103805-488EA
21
USD
103805-488
103843-726
77150-900
77150-902
77150-904
77300-534
77300-536
77319-704
4-Bromo-5-chloropyridin-2-amine 95%
4-Bromo-5-chloropyridin-2-amine
Precaución: for Research Use Only
Formula:
C₅H₄BrClN₂ MW: 207.46 g/mol Boiling Pt: 299.8±35.0 °C (760 mm Hg) Flash Pt: 135.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD15143360 CAS Number: 1187449-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...