4-Bromo-5-chloro-2-fluorobenzaldehyde 95%

Supplier: Ambeed
Warning

A135513-1G A135513-5G A135513-250MG
103802-164EA 108 USD
103802-164 77314-996 77314-998 77685-168
4-Bromo-5-chloro-2-fluorobenzaldehyde 95%
4-Bromo-5-chloro-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrClFO
Storage Temperature: Refrigerator
MDL Number: MFCD26142710
CAS Number: 1603584-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR