4-Bromo-4'-methylbiphenyl 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-4'-methyl-1,1'-biphenyl

A101838-1G A101838-5G
103796-168EA 10 USD
103796-168 103821-862 77308-754 77308-756
4-Bromo-4'-methylbiphenyl 97%
4-Bromo-4'-methylbiphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₁Br
MW: 247.14 g/mol
Boiling Pt: 322.9 °C (760 mm Hg)
Melting Pt: 136 °C
Flash Pt: 148.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00466236
CAS Number: 50670-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR