4-Bromo-4'-iodobiphenyl 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-4'-iodo-1,1'-biphenyl, 1-Bromo-4-(4-iodophenyl)benzene
A385597-25G
A385597-5G
A385597-10G
A385597-100G
103837-826EA
5
USD
103837-826
76841-168
76841-170
76876-728
77568-776
4-Bromo-4'-iodobiphenyl 98%
4-Bromo-4'-iodobiphenyl
Precaución: For research use only.
Formula:
C₁₂H₈BrI MW: 359.00 g/mol Boiling Pt: 369.3 °C (760 mm Hg) Melting Pt: 166…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD17676204 CAS Number: 105946-82-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...