4-Bromo-3-nitrobenzonitrile 97%

Supplier: Ambeed
Warning

A296352-5G A296352-100G A296352-10G
103814-092EA 6 USD
103814-092 103835-232 76837-898 77114-394 77114-396
4-Bromo-3-nitrobenzonitrile 97%
4-Bromo-3-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrN₂O₂
Boiling Pt: 268.6 °C (760 mm Hg)
Melting Pt: 116…120 °C
Flash Pt: 116.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00982006
CAS Number: 89642-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR