4-Bromo-m-toluonitrile 98%

Supplier: Ambeed
Warning

A133996-100G A133996-10G A133996-500G A133996-1G A133996-25G A133996-5G
103801-950EA 5 USD
103801-950 77234-640 77234-642 77234-644 77298-674 77314-656 77314-658
4-Bromo-m-toluonitrile 98%
4-Bromo-m-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 265 °C (760 mm Hg)
Melting Pt: 53…57 °C
Flash Pt: 96.11 °C
Storage Temperature: Ambient
MDL Number: MFCD00031538
CAS Number: 41963-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR