4-Bromo-m-toluic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-3-methylbenzoic acid, 2-Bromo-5-carboxytoluene, 4-Bromo-m-toluic acid

A162815-100G A162815-10G A162815-1G A162815-500G A162815-25G A162815-5G
103843-912EA 6 USD
103843-912 76689-880 76689-882 76689-884 76689-886 77300-608 77319-986
4-Bromo-m-toluic acid 98%
4-Bromo-m-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Melting Pt: 212…216 °C
Flash Pt: 142.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00039526
CAS Number: 7697-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR