4-Bromo-3-methylbenzaldehyde 98%

Supplier: Ambeed
Warning

A133103-100G A133103-10G A133103-500G A133103-25G A133103-1G A133103-5G
103801-826EA 6 USD
103801-826 77285-380 77285-382 77285-384 77298-608 77314-428 77314-430
4-Bromo-3-methylbenzaldehyde 98%
4-Bromo-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO
Boiling Pt: 262 °C (760 mm Hg)
Flash Pt: 96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07787170
CAS Number: 78775-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR