4-Bromo-m-toluidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Amino-6-bromotoluene, 4-Bromo-3-methylaniline, 5-Amino-2-bromotoluene
A386475-100G
A386475-10G
A386475-500G
A386475-25G
A386475-5G
103837-840EA
6
USD
103837-840
76817-718
76817-720
76817-722
76841-196
77306-974
4-Bromo-m-toluidine 98%
4-Bromo-m-toluidine
Precaución: for Research Use Only
Formula:
C₇H₈BrN MW: 186.05 g/mol Boiling Pt: 240 °C (760 mm Hg) Melting Pt: 79…83 °C Flash Pt: 101.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007828 CAS Number: 6933-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...