4-Bromo-3-methyl-1H-pyrrolo[2,3-b]pyridine 97%
Supplier: Ambeed
|
Warning
|
A149096-100MG
A149096-250MG
A149096-10G
103804-000EA
128
USD
103804-000
103842-120
77162-692
77162-694
77892-025
4-Bromo-3-methyl-1H-pyrrolo[2,3-b]pyridine 97%
4-Bromo-3-methyl-1H-pyrrolo[2,3-b]pyridine
Precaución: for Research Use Only
Formula:
C₈H₇BrN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD22566175 CAS Number: 1363382-02-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...