4-Bromo-3-methoxyphenol 98%

Supplier: Ambeed
Warning

A342747-1G A342747-25G A342747-5G A342747-10G A342747-250MG
103825-870EA 12 USD
103825-870 103836-844 76839-892 76839-894 76839-896 77290-548 77290-550
4-Bromo-3-methoxyphenol 98%
4-Bromo-3-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇BrO₂
Melting Pt: 77…82 °C
Flash Pt: 122.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00029748
CAS Number: 102127-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR