4-Bromo-3-methoxybenzonitrile 98%

Supplier: Ambeed
Warning

A355835-1G A355835-25G A355835-5G A355835-10G A355835-100G
103826-152EA 7 USD
103826-152 103837-160 76840-290 76840-292 76840-294 77290-690 77748-705
4-Bromo-3-methoxybenzonitrile 98%
4-Bromo-3-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrNO
MW: 212.05 g/mol
Boiling Pt: 265.3 °C (760 mm Hg)
Flash Pt: 114.25 °C
Storage Temperature: Ambient
MDL Number: MFCD09753723
CAS Number: 120315-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR