4-Bromo-3-methoxybenzaldehyde 97%

Supplier: Ambeed
Warning

A131965-100G A131965-10G A131965-250MG A131965-1G A131965-5G
103801-666EA 15 USD
103801-666 76972-118 76972-120 76972-122 77314-200 77314-202
4-Bromo-3-methoxybenzaldehyde 97%
4-Bromo-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
Melting Pt: 126…127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11846303
CAS Number: 43192-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR