4-Bromo-m-anisidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-3-methoxyaniline, 5-Amino-2-bromoanisole, 4-Bromo-3-anisidine

A298192-5G A298192-100G A298192-10G A298192-500G A298192-1G A298192-25G
103814-178EA 9 USD
103814-178 103835-322 76838-018 77014-892 77014-894 77014-896 77305-738 77305-740
4-Bromo-m-anisidine 98%
4-Bromo-m-anisidine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.05 g/mol
Boiling Pt: 272.1 °C (760 mm Hg)
Melting Pt: 96…100 °C
Flash Pt: 118.3 °C
Storage Temperature: Ambient
MDL Number: MFCD05664063
CAS Number: 19056-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR