4-Bromo-3-iodonitrobenzene 95%

Supplier: Ambeed
Warning

A306924-1G A306924-25G A306924-100MG A306924-250MG
103814-488EA 38 USD
103814-488 103835-660 76838-394 76838-396 77099-652 77099-654
4-Bromo-3-iodonitrobenzene 95%
4-Bromo-3-iodonitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrINO₂
Boiling Pt: 330.6 °C (760 mm Hg)
Flash Pt: 153.8 °C
Storage Temperature: Ambient
MDL Number: MFCD17012734
CAS Number: 63037-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR