4-Bromo-3-hydroxybenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A236076-25G
A236076-5G
A236076-100G
A236076-100MG
A236076-10G
A236076-500G
A236076-1G
103811-070EA
6
USD
103811-070
103850-840
77089-880
77089-882
77288-570
77288-572
77288-574
77288-576
77304-030
4-Bromo-3-hydroxybenzoic acid 98%
4-Bromo-3-hydroxybenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₅BrO₃ Boiling Pt: 338.6 °C (760 mm Hg) Melting Pt: 225…227 °C Flash Pt: 158.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD08056371 CAS Number: 14348-38-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...