4-Bromo-3-formylbenzonitrile 95%

Supplier: Ambeed
Warning

A200255-100MG A200255-250MG A200255-1G A200255-5G A200255-10G A200255-25G
103817-180EA 29 USD
103817-180 103847-808 76814-260 76814-262 77085-866 77085-868 77883-453 77883-454
4-Bromo-3-formylbenzonitrile 95%
4-Bromo-3-formylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₄BrNO
Boiling Pt: 253.927 °C (760 mm Hg)
Melting Pt: 123…127 °C
Flash Pt: 107.372 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11858309
CAS Number: 89003-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR