4-Bromo-3-fluorobenzonitrile 98%

Supplier: Ambeed
Warning

A107994-100G A107994-10G A107994-1G A107994-25G A107994-500G A107994-5G
103823-050EA 9 USD
103823-050 76765-610 76765-612 76765-614 76765-616 76765-618 77283-672
4-Bromo-3-fluorobenzonitrile 98%
4-Bromo-3-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrFN
Boiling Pt: 230.1 °C (760 mm Hg)
Melting Pt: 54…58 °C
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD07782069
CAS Number: 133059-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR