4-Bromo-3-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

A141068-250MG A141068-1G A141068-5G
103802-946EA 39 USD
103802-946 77095-444 77299-194 77299-196
4-Bromo-3-fluorophenylboronic acid 97%
4-Bromo-3-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrFO₂
Boiling Pt: 314.6 °C (760 mm Hg)
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093066
CAS Number: 374790-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR