4-Bromo-3-fluoro-2-methylpyridine 95%

Supplier: Ambeed
Warning

A228970-250MG A228970-5G A228970-1G A228970-100MG
103819-432EA 171 USD
103819-432 76987-266 76987-268 77089-150 77612-226
4-Bromo-3-fluoro-2-methylpyridine 95%
4-Bromo-3-fluoro-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFN
MW: 190.013 g/mol
Boiling Pt: 181.2±35.0 °C (760 mm Hg)
Flash Pt: 63.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16609967
CAS Number: 1211583-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR