4-Bromo-3-chloropyridine 95%

Supplier: Ambeed
Warning

A226007-100G A226007-250MG
103819-214EA 9 USD
103819-214 103850-116 76874-112 76874-114
4-Bromo-3-chloropyridine 95%
4-Bromo-3-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrClN
Boiling Pt: 219.8 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD04112577
CAS Number: 73583-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR