4-Bromo-3-chlorophenol 98%

Supplier: Ambeed
Warning

A142801-100G A142801-10G A142801-1G A142801-25G A142801-5G
103803-214EA 5 USD
103803-214 77285-820 77285-822 77316-458 77316-460 77316-462
4-Bromo-3-chlorophenol 98%
4-Bromo-3-chlorophenol

Precaución: for Research Use Only

Formula: C₆H₄BrClO
MW: 207.45 g/mol
Boiling Pt: 267 °C (760 mm Hg)
Melting Pt: 64…66 °C
Flash Pt: 115.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00070744
CAS Number: 13631-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR