4-Bromo-3-chlorobenzonitrile 98%

Supplier: Ambeed
Warning

A109117-100G A109117-10G A109117-250MG A109117-25G A109117-1G A109117-5G
103797-448EA 6 USD
103797-448 103823-238 76970-602 76970-604 76970-606 76970-608 77310-054 77310-056
4-Bromo-3-chlorobenzonitrile 98%
4-Bromo-3-chlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrClN
MW: 216.46 g/mol
Boiling Pt: 261 °C (760 mm Hg)
Melting Pt: 80…81 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD09834777
CAS Number: 57418-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR