4-Bromo-3-chlorobenzaldehyde 98%

Supplier: Ambeed
Warning

A147800-10G A147800-1G A147800-25G A147800-5G A147800-100G
103803-836EA 11 USD
103803-836 103841-944 76783-236 77317-348 77317-350 77317-352 77667-100
4-Bromo-3-chlorobenzaldehyde 98%
4-Bromo-3-chlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrClO
MW: 219.4600 g/mol
Boiling Pt: 281.3 °C (760 mm Hg)
Flash Pt: 123.899 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08059110
CAS Number: 120077-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR