4-Bromo-3,6-dichloropyridazine 95%

Supplier: Ambeed
Warning

A119794-100MG A119794-250MG A119794-1G A119794-5G
103799-840EA 24 USD
103799-840 76810-412 76810-414 77311-944 77311-946
4-Bromo-3,6-dichloropyridazine 95%
4-Bromo-3,6-dichloropyridazine

Precaución: for Research Use Only

Formula: C₄HBrCl₂N₂
MW: 227.87 g/mol
Boiling Pt: 341.8 °C (760 mm Hg)
Flash Pt: 160.512 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13193322
CAS Number: 10344-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR