4-Bromo-3,5-xylenol 98%

Supplier: Ambeed
Warning

A246250-1000G A246250-100G A246250-10G A246250-500G A246250-25G
103851-528EA 6 USD
103851-528 77249-882 77249-884 77249-886 77249-888 77304-380
4-Bromo-3,5-xylenol 98%
4-Bromo-3,5-xylenol

Precaución: for Research Use Only

Formula: C₈H₉BrO
Melting Pt: 113…115 °C
Flash Pt: 115.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00002315
CAS Number: 7463-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR