4-Bromo-3,5-dimethylbenzonitrile 98%
Supplier: Ambeed
|
Warning
|
A140149-250MG
A140149-1G
A140149-25G
A140149-5G
A140149-100G
103802-836EA
13
USD
103802-836
77109-768
77109-770
77315-944
77315-946
77315-948
77685-198
4-Bromo-3,5-dimethylbenzonitrile 98%
4-Bromo-3,5-dimethylbenzonitrile
Precaución: for Research Use Only
Formula:
C₉H₈BrN Boiling Pt: 291 °C (760 mm Hg) Melting Pt: 135…137 °C Flash Pt: 130 °C Storage Temperature: Ambient |
MDL Number:
MFCD07369910 CAS Number: 75344-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...