4-Bromo-3,5-dimethylbenzaldehyde 98%

Supplier: Ambeed
Warning

A163297-10G A163297-250MG A163297-1G
103805-710EA 30 USD
103805-710 103843-970 77259-714 77259-716 77320-058
4-Bromo-3,5-dimethylbenzaldehyde 98%
4-Bromo-3,5-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO
MW: 213.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08063822
CAS Number: 400822-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR