4-Bromo-3,5-xylidine 98%

Supplier: Ambeed
Warning

A234078-1G A234078-5G A234078-10G A234078-25G A234078-500G A234078-100G
103810-926EA 6 USD
103810-926 103850-686 77089-658 77089-660 77288-490 77288-492 77288-494 77748-403
4-Bromo-3,5-xylidine 98%
4-Bromo-3,5-xylidine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
Boiling Pt: 261 °C (760 mm Hg)
Melting Pt: 73…74 °C
Flash Pt: 111.644 °C
Storage Temperature: Ambient
MDL Number: MFCD07780648
CAS Number: 59557-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR