4-Bromo-3,5-dimethoxypyridine 98%

Supplier: Ambeed
Warning

A160747-100MG A160747-250MG
103805-414EA 90 USD
103805-414 103843-648 76812-716 76812-718
4-Bromo-3,5-dimethoxypyridine 98%
4-Bromo-3,5-dimethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO₂
MW: 218.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14705143
CAS Number: 1033610-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR