4-Bromo-3,5-difluorobenzoic acid 95%

Supplier: Ambeed
Warning

A147437-100G A147437-100MG A147437-10G A147437-250MG A147437-1G A147437-25G A147437-5G
103803-778EA 15 USD
103803-778 103841-880 76823-690 76823-692 76823-694 76823-696 77317-250 77317-252 77317-254
4-Bromo-3,5-difluorobenzoic acid 95%
4-Bromo-3,5-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O₂
MW: 237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917156
CAS Number: 651027-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR