4-Bromo-3,5-dichlorobenzoic acid 98%

Supplier: Ambeed
Warning

A163901-100G A163901-10G A163901-250MG A163901-1G A163901-25G A163901-5G
103805-786EA 21 USD
103805-786 103844-050 76824-188 76824-190 76824-192 77300-680 77320-172 77320-174
4-Bromo-3,5-dichlorobenzoic acid 98%
4-Bromo-3,5-dichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrCl₂O₂
MW: 269.91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14704247
CAS Number: 117738-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR