4-Bromo-3,5-bis(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

A261942-1G A261942-10G A261942-250MG
103812-550EA 45 USD
103812-550 103852-442 77092-060 77193-850 77193-852
4-Bromo-3,5-bis(trifluoromethyl)aniline 98%
4-Bromo-3,5-bis(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₈H₄BrF₆N
MW: 308.0185 g/mol
Boiling Pt: 210.7 °C (760 mm Hg)
Melting Pt: 78…80 °C
Flash Pt: 81.2 °C
Storage Temperature: Ambient
MDL Number: MFCD04973750
CAS Number: 268733-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR