4-Bromo-3-(trifluoromethyl)benzoic acid 97%

Supplier: Ambeed
Warning

A113319-100G A113319-10G A113319-25G A113319-1G A113319-5G
103798-216EA 9 USD
103798-216 103824-056 77108-108 77108-110 77108-112 77310-870 77310-872
4-Bromo-3-(trifluoromethyl)benzoic acid 97%
4-Bromo-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₂
Boiling Pt: 289.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08459288
CAS Number: 161622-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR