4-Bromo-3-(trifluoromethyl)-1H-pyrazole 97%

Supplier: Ambeed
Warning

A152396-10G A152396-250MG
103804-388EA 14 USD
103804-388 103842-556 77890-000 77890-001
4-Bromo-3-(trifluoromethyl)-1H-pyrazole 97%
4-Bromo-3-(trifluoromethyl)-1H-pyrazole

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₄H₂BrF₃N₂
Boiling Pt: 235.3 °C (760 mm Hg)
Melting Pt: 105…107 °C
Flash Pt: 96.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04969230
CAS Number: 19968-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR