4-Bromo-3-(hydroxymethyl)benzoic acid 95%

Supplier: Ambeed
Warning

A158914-250MG A158914-1G A158914-5G
103805-190EA 89 USD
103805-190 103843-402 76984-974 77319-260 77319-262
4-Bromo-3-(hydroxymethyl)benzoic acid 95%
4-Bromo-3-(hydroxymethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
MW: 231.043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16249537
CAS Number: 790230-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR