4-Bromo-2-vinylpyridine 95%

Supplier: Ambeed
Warning

A257493-1MG A257493-250MG
103812-306EA 163 USD
103812-306 103852-174 76815-912 76815-914
4-Bromo-2-vinylpyridine 95%
4-Bromo-2-vinylpyridine

Precaución: for Research Use Only

Formula: C₇H₆BrN
MW: 184.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09833154
CAS Number: 502509-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR