4-Bromo-2-thenoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-thiophenecarboxylic acid

A141112-100G A141112-10G A141112-1G A141112-25G A141112-5G
103802-954EA 9 USD
103802-954 77285-748 77285-750 77316-080 77316-082 77316-084
4-Bromo-2-thenoic acid 98%
4-Bromo-2-thenoic acid

Precaución: for Research Use Only

Formula: C₅H₃BrO₂S
MW: 207.05 g/mol
Boiling Pt: 323.7 °C (760 mm Hg)
Melting Pt: 121 °C
Flash Pt: 149.594 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03422294
CAS Number: 16694-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR