4-Bromo-2-nitropyridine 97%

Supplier: Ambeed
Warning

A172953-100MG A172953-250MG A172953-1G A172953-5G
103806-770EA 206 USD
103806-770 103845-062 76985-528 76985-530 77082-284 77082-286
4-Bromo-2-nitropyridine 97%
4-Bromo-2-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrN₂O₂
MW: 202.99 g/mol
Boiling Pt: 305.1±22.0 °C (760 mm Hg)
Flash Pt: 138.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD26743706
CAS Number: 909712-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR