4-Bromo-2-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A140952-100G A140952-10G A140952-500G A140952-25G A140952-5G
76767-776EA 142.17 USD
76767-776 76767-778 76767-780 77316-046 77316-048
4-Bromo-2-nitrobenzoic acid 97%
4-Bromo-2-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrNO₄
MW: 246.02 g/mol
Melting Pt: 165…169 °C
Flash Pt: 176.7 °C
Storage Temperature: Ambient
MDL Number: MFCD01013599
CAS Number: 99277-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR