4-Bromo-2-nitrobenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A255650-100G
A255650-10G
A255650-500G
A255650-1G
A255650-25G
A255650-5G
103812-200EA
5
USD
103812-200
103852-062
76647-774
76647-776
76647-778
77091-520
77304-654
77304-656
4-Bromo-2-nitrobenzaldehyde 97%
4-Bromo-2-nitrobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₄BrNO₃ MW: 230.0156 g/mol Boiling Pt: 334.4 °C (760 mm Hg) Flash Pt: 156.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00463687 CAS Number: 5551-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...