4-Bromo-2-nitroaniline 98%

Supplier: Ambeed
Warning

A255531-25G A255531-5G A255531-100G A255531-10G A255531-1G A255531-500G
103852-056EA 6 USD
103852-056 77091-514 77091-516 77098-736 77098-738 77098-740 77098-742
4-Bromo-2-nitroaniline 98%
4-Bromo-2-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00041312
CAS Number: 875-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR